missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,5-Tetrafluorobenzoyl chloride, 99%, ACROS Organics™

Chemical Identifiers

CAS 94695-48-4
Molecular Formula C7HClF4O
Molecular Weight (g/mol) 212.53
MDL Number MFCD00075164
InChI Key XWCKIXLTBNGIHV-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro benzoyl chloride, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobenzoylchloride, 2,3,4,5-tetrafluorobezoyl chloride, 2,3,4,5-tetrafluorobezoylchloride, benzoyl chloride, 2,3,4,5-tetrafluoro, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, tetrafluorobenzoyl chloride
PubChem CID 2733689
IUPAC Name 2,3,4,5-tetrafluorobenzoyl chloride
SMILES C1=C(C(=C(C(=C1F)F)F)F)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334570010
SDS View Documents
Thermo Fisher Scientific
334570010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

94695-48-4
212.53
XWCKIXLTBNGIHV-UHFFFAOYSA-N
2733689
C1=C(C(=C(C(=C1F)F)F)F)C(=O)Cl
C7HClF4O
MFCD00075164
2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro benzoyl chloride, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobenzoylchloride, 2,3,4,5-tetrafluorobezoyl chloride, 2,3,4,5-tetrafluorobezoylchloride, benzoyl chloride, 2,3,4,5-tetrafluoro, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, tetrafluorobenzoyl chloride
2,3,4,5-tetrafluorobenzoyl chloride

Specifications

1.5800g/mL
Authentic
1.4780 to 1.49
Colorless
94695-48-4
100.0
C7HClF4O
2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro benzoyl chloride, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobenzoylchloride, 2,3,4,5-tetrafluorobezoyl chloride, 2,3,4,5-tetrafluorobezoylchloride, benzoyl chloride, 2,3,4,5-tetrafluoro, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, tetrafluorobenzoyl chloride
C1=C(C(=C(C(=C1F)F)F)F)C(=O)Cl
212.53
212.53
99%
90°C
Glass bottle
1.58
1g
98.5
98.5% min. (GC)
MFCD00075164
XWCKIXLTBNGIHV-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzoyl chloride
2733689
Liquid
2,3,4,5-Tetrafluorobenzoyl chloride, 99%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

TSCA : TSCA

SDS