Please login to your online account to display your discounted pricing

2,3,4,5-Tetrafluorobenzotrifluoride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,4,5-Tetrafluorobenzotrifluoride
Name Note 99%
CAS 654-53-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7 H F7
Formula Weight 218.07
MDL Number MFCD01862013
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334580050

  SDS
thermo fisher scientific
334580050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC334580010

  SDS
thermo fisher scientific
334580010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 218.07
Quantity 5g
Color Clear colorless
TSCA TSCA
CAS Max % 100.0
Refractive Index 1.3715 to 1.3735 (20°C, 589nm)
Assay Percent Range 99%
MDL Number MFCD01862013
CAS 654-53-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2,3,4,5-Tetrafluorobenzotrifluoride
Packaging Glass bottle
Name Note 99%
Molecular Formula C7 H F7
Synonym α,α,α,2,3,4,5-Heptafluorotoluene
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.