Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2,3,4,5-Tetrafluorobenzonitrile
CAS 16582-93-7
Molecular Formula C7HF4N
Formula Weight 175.09
MDL Number MFCD00040187
UN Number UN3276
Beilstein 3092489
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2281306

  SDS
alfa aesar™
B2281306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2281314

  SDS
alfa aesar™
B2281314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2281322

  SDS
alfa aesar™
B2281322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.09
Chemical Name or Material 2,3,4,5-Tetrafluorobenzonitrile
Assay 97%
Molecular Formula C7HF4N
MDL Number MFCD00040187
CAS 16582-93-7
UN Number UN3276
Beilstein 3092489
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 164° to 165°C
Flash Point 70°C (158°F)
Odor Characteristic
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.