Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzoic acid, 98+%

$21.70 - $21.70

Chemical Identifiers

Chemical Name or Material 2,3,4,5-Tetrafluorobenzoic acid
CAS 1201-31-6
Molecular Formula C7H2F4O2
Formula Weight 194.09
MDL Number MFCD00009613
Beilstein 2052670
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1895403

  SDS
alfa aesar™
A1895403
1g Each for $21.70
Description & Specifications

Specifications

Formula Weight 194.09
Quantity 1g
Beilstein 2052670
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H2F4O2
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1201-31-6
Assay ≥98%
MDL Number MFCD00009613
Health Hazard 1 Warning
Melting Point 85° to 87°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.