Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzeneboronic acid, 97%

$363.27 - $363.27

Chemical Identifiers

Chemical Name or Material 2,3,4,5-Tetrafluorobenzeneboronic acid
CAS 179923-32-1
Molecular Formula C6H3BF4O2
Formula Weight 193.9
MDL Number MFCD04039308
Beilstein 8554485
Synonym 2,3,4,5-Tetrafluorophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1982403

 
alfa aesar™
L1982403
1g Each for $363.27
Description & Specifications

Specifications

Formula Weight 193.9
Chemical Name or Material 2,3,4,5-Tetrafluorobenzeneboronic acid
Molecular Formula C6H3BF4O2
MDL Number MFCD04039308
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
CAS 179923-32-1
Synonym 2,3,4,5-Tetrafluorophenylboronic acid
Health Hazard 1 Warning
Melting Point ≈250° to 255°C
Quantity 1g
Beilstein 8554485
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.