Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzamide, 97+%

$209.45 - $209.45

Chemical Identifiers

Chemical Name or Material 2,3,4,5-Tetrafluorobenzamide
CAS 16582-94-8
Molecular Formula C7H3F4NO
Formula Weight 193.1
MDL Number MFCD01631457
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2295806

  SDS
alfa aesar™
B2295806
5g Each for $209.45
Description & Specifications

Specifications

Formula Weight 193.1
Chemical Name or Material 2,3,4,5-Tetrafluorobenzamide
CAS 16582-94-8
Quantity 5g
Melting Point 132° to 134°C
MDL Number MFCD01631457
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥97%
Molecular Formula C7H3F4NO
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.