missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3:4,5-Di-O-isopropylidene-D-arabitol, 98%

$108.41 - $191.22

Chemical Identifiers

CAS 19139-74-3
Molecular Formula C11H20O5
Molecular Weight (g/mol) 232.276
MDL Number MFCD00063387
InChI Key FBIWGPWXTDOFQR-HRDYMLBCSA-N
Synonym 2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
PubChem CID 11042611
IUPAC Name [(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(OCC(O1)C2C(OC(O2)(C)C)CO)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66608ME View Documents Alfa Aesar™
J66608ME
500mg Each for $108.41
 
AAJ6660803 View Documents Alfa Aesar™
J6660803
1g Each for $191.22
 
Description

Description

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
Specifications

Chemical Identifiers

19139-74-3
232.276
FBIWGPWXTDOFQR-HRDYMLBCSA-N
11042611
CC1(OCC(O1)C2C(OC(O2)(C)C)CO)C
C11H20O5
MFCD00063387
2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
[(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol

Specifications

Colorless
19139-74-3
MFCD00063387
FBIWGPWXTDOFQR-HRDYMLBCSA-N
[(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
11042611
Viscous Liquid
2,3:4,5-Di-O-isopropylidene-D-arabitol
500mg
C11H20O5
2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
CC1(OCC(O1)C2C(OC(O2)(C)C)CO)C
232.276
232.27
98%
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C