Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentamethylbenzyl alcohol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol
CAS 484-66-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12 H18 O
Linear Formula C6(CH3)5CH2OH
Formula Weight 178.27
MDL Number MFCD00014421
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372780010

  SDS
thermo fisher scientific
AC372780010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 178.27
Physical Form Crystalline Powder
Quantity 1g
Color White
Melting Point 159° to 163°C
CAS Max % 100.0
Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C12 H18 O
Linear Formula C6(CH3)5CH2OH
MDL Number MFCD00014421
CAS 484-66-2
CAS Min % 97.5
Infrared Spectrum Authentic