Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentamethylbenzyl alcohol, 96%

$25.47 - $247.11

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol
CAS 484-66-2
Molecular Formula C12H18O
Formula Weight 178.28
MDL Number MFCD00014421
Beilstein 2208765
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2257403

  SDS
alfa aesar™
B2257403
1g Each for $25.47

AAB2257406

  SDS
alfa aesar™
B2257406
5g Each for $84.82

AAB2257414

  SDS
alfa aesar™
B2257414
25g Each for $247.11
Description & Specifications

Specifications

Formula Weight 178.28
Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol
Assay 96%
Melting Point 158° to 160°C
Molecular Formula C12H18O
Quantity 1g
MDL Number MFCD00014421
CAS 484-66-2
Beilstein 2208765