Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentamethylbenzophenone, 98%

$80.04 - $363.27

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentamethylbenzophenone
CAS 20386-33-8
Molecular Formula C18H20O
Formula Weight 252.36
MDL Number MFCD00051595
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1518206

 
alfa aesar™
A1518206
5g Each for $80.04

AAA1518214

 
alfa aesar™
A1518214
25g Each for $363.27
Description & Specifications

Specifications

Formula Weight 252.36
Chemical Name or Material 2,3,4,5,6-Pentamethylbenzophenone
Assay 98%
Melting Point 137° to 138°C
Molecular Formula C18H20O
Quantity 5g
MDL Number MFCD00051595
CAS 20386-33-8