Alfa Aesar™ 2,3,4,5,6-Pentamethylbenzophenone, 98%
Manufacturer:  Alfa Aesar™ A1518206
 View more versions of this product
Catalog No. AAA1518206
Specifications
Specifications
2,3,4,5,6-Pentamethylbenzophenone
5g
C18H20O
2,3,4,5,6-pentamethylbenzophenone, 2,3,4,5,6-pentamethylphenyl phenyl methanone, pentamethylphenyl phenyl methanone, 2,3,4,5,6-pentamethyl benzopheneone, acmc-20aoiy, maybridge1_002590, methanone, pentamethylphenyl phenyl-9ci, 2,3,4,5,6-pentamethylphenyl phenyl ketone, 2,3,4,5,6-pentamethylphenyl-phenylmethanone, 2,3,4,5,6-pentamethylphenyl phenyl methanone #
CC1=C(C(=C(C(=C1C)C)C(=O)C2=CC=CC=C2)C)C
252.357
252.36
137°C to 138°C
20386-33-8
MFCD00051595
JFFVBXWGARSRSC-UHFFFAOYSA-N
(2,3,4,5,6-pentamethylphenyl)-phenylmethanone
519768
98%
Chemical Identifiers
20386-33-8
252.357
JFFVBXWGARSRSC-UHFFFAOYSA-N
519768
CC1=C(C(=C(C(=C1C)C)C(=O)C2=CC=CC=C2)C)C
C18H20O
MFCD00051595
2,3,4,5,6-pentamethylbenzophenone, 2,3,4,5,6-pentamethylphenyl phenyl methanone, pentamethylphenyl phenyl methanone, 2,3,4,5,6-pentamethyl benzopheneone, acmc-20aoiy, maybridge1_002590, methanone, pentamethylphenyl phenyl-9ci, 2,3,4,5,6-pentamethylphenyl phenyl ketone, 2,3,4,5,6-pentamethylphenyl-phenylmethanone, 2,3,4,5,6-pentamethylphenyl phenyl methanone #
(2,3,4,5,6-pentamethylphenyl)-phenylmethanone
Documents