Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3',4',5',6'-Pentamethylacetophenone, 98+%

$51.38 - $485.89

Chemical Identifiers

Chemical Name or Material 2',3',4',5',6'-Pentamethylacetophenone
CAS 2040-01-9
Molecular Formula C13H18O
Formula Weight 190.29
MDL Number MFCD00014986
Beilstein 1867535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1904006

  SDS
alfa aesar™
A1904006
5g Each for $51.38

AAA1904022

  SDS
alfa aesar™
A1904022
100g Each for $485.89
Description & Specifications

Specifications

Formula Weight 190.29
Chemical Name or Material 2',3',4',5',6'-Pentamethylacetophenone
Assay ≥98%
Melting Point 84° to 87°C
Molecular Formula C13H18O
Quantity 5g
MDL Number MFCD00014986
CAS 2040-01-9
Beilstein 1867535