Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentafluorophenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile
CAS 653-30-5
Molecular Formula C8H2F5N
Formula Weight 207.1
MDL Number MFCD00013814
UN Number UN3276
Beilstein 1882341
Synonym 2,3,4,5,6-Pentafluorobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2223903

  SDS
alfa aesar™
B2223903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2223906

  SDS
alfa aesar™
B2223906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.1
Health Hazard 1 Danger
Molecular Formula C8H2F5N
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile
Beilstein 1882341
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Quantity 1g
Synonym 2,3,4,5,6-Pentafluorobenzyl cyanide
Boiling Point 110° to 111°C (17mmHg)
Flash Point >110°C (230°F)
CAS 653-30-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3276
Assay 98%
MDL Number MFCD00013814

Applications
2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.