missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentafluorophenylacetonitrile, 96%, ACROS Organics™

Chemical Identifiers

CAS 653-30-5
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C8H2F5N
Linear Formula C6F5CH2CN
Formula Weight 207.1
MDL Number MFCD00013814
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile, 96%
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC296160010 SDS Thermo Fisher Scientific
AC296160010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

653-30-5
100.0
C8H2F5N
207.1
2,3,4,5,6-Pentafluorophenylacetonitrile, 96%
95.0
96%
C6F5CH2CN
MFCD00013814
≥95%

Specifications

1.6100g/mL
1g
95.0
Authentic
1.4390 to 1.4410 (20°C, 589nm)
96%
C6F5CH2CN
MFCD00013814
≥95%
Clear colorless to slightly yellow
653-30-5
100.0
Glass bottle
Liquid
C8H2F5N
207.1
2,3,4,5,6-Pentafluorophenylacetonitrile, 96%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-498-6

TSCA : TSCA