Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentafluorophenoxyacetic acid, 98+%

$27.12 - $99.89

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentafluorophenoxyacetic acid
CAS 14892-14-9
Molecular Formula C8H3F5O3
Formula Weight 242.1
MDL Number MFCD00004297
Beilstein 1888139
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2273103

  SDS
alfa aesar™
B2273103
1g Each for $27.12

AAB2273106

  SDS
alfa aesar™
B2273106
5g Each for $99.89
Description & Specifications

Specifications

Formula Weight 242.1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 108° to 110°C
Assay ≥98%
CAS 14892-14-9
Molecular Formula C8H3F5O3
Health Hazard 1 Warning
Beilstein 1888139
Quantity 1g
MDL Number MFCD00004297
Chemical Name or Material 2,3,4,5,6-Pentafluorophenoxyacetic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.