Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentafluorocinnamic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentafluorocinnamic acid
Name Note 98%
CAS 719-60-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H3F5O2
Linear Formula C6F5CH=CHCO2H
Formula Weight 238.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC365350050

  SDS
thermo fisher scientific
AC365350050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 238.11
Quantity 5g
Color White to yellow
Melting Point 150° to 154°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula C6F5CH=CHCO2H
Molecular Formula C9H3F5O2
Assay Percent Range 98%
MDL Number MFCD00004371
CAS Min % 97.5
Name Note 98%
CAS 719-60-8
Chemical Name or Material 2,3,4,5,6-Pentafluorocinnamic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.