Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzyl chloride, 98%

$118.58 - $118.58

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl chloride
CAS 653-35-0
Molecular Formula C7H2ClF5
Formula Weight 216.54
MDL Number MFCD00040864
UN Number UN2922
Beilstein 647807
Synonym alpha-Chloro-2,3,4,5,6-pentafluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1268706

  SDS
alfa aesar™
L1268706
5g Each for $118.58

AAL1268703

  SDS
alfa aesar™
L1268703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.54
CAS 653-35-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Odor Pungent
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl chloride
Assay 98%
Boiling Point 72°C (22mmHg)
Molecular Formula C7H2ClF5
Quantity 5g
MDL Number MFCD00040864
UN Number UN2922
Beilstein 647807
Synonym alpha-Chloro-2,3,4,5,6-pentafluorotoluene
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Applications
2,3,4,5,6-Pentafluorobenzyl chloride is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, alkali metals and alkaline earth metals.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.