missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzyl alcohol, 98%

Manufacturer:  Alfa Aesar™ A1211922

 View more versions of this product

Catalog No. AAA1211922



Specifications

Specifications

2,3,4,5,6-Pentafluorobenzyl alcohol
98%
MFCD00004602
PGJYYCIOYBZTPU-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl)methanol
9923
198.09
440-60-8
C7H3F5O
2052669
C(C1=C(C(=C(C(=C1F)F)F)F)F)O
198.092
CHEBI:44903

Chemical Identifiers

440-60-8
198.092
PGJYYCIOYBZTPU-UHFFFAOYSA-N
CHEBI:44903
C(C1=C(C(=C(C(=C1F)F)F)F)F)O
C7H3F5O
MFCD00004602
9923
(2,3,4,5,6-pentafluorophenyl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DP0695000

SDS Documents