Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentafluorobenzyl alcohol, 96-98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  188360250

 View more versions of this product

Catalog No. AC188360250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Mass
Assay ≥96%
Formula Weight 198.09
Quantity 25g
Color White
Melting Point 34° to 39°C
CAS Max % 100.0
CAS Min % 96.0
Assay Percent Range 96-98%
Molecular Formula C7H3F5O
MDL Number MFCD00004602
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol
Packaging Glass bottle
Linear Formula C6F5CH2OH
CAS 440-60-8
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol
CAS 440-60-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 96-98%
Molecular Formula C7H3F5O
Linear Formula C6F5CH2OH
Formula Weight 198.09
MDL Number MFCD00004602
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents