Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzophenone, 98%

$74.45 - $1,167.74

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentafluorobenzophenone
CAS 1536-23-8
Molecular Formula C13H5F5O
Formula Weight 272.18
MDL Number MFCD00000294
Beilstein 2059098
Synonym Pentafluorophenyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1371306

  SDS
alfa aesar™
A1371306
5g Each for $74.45

AAA1371314

  SDS
alfa aesar™
A1371314
25g Each for $317.93

AAA1371322

  SDS
alfa aesar™
A1371322
100g Each for $1,167.74
Description & Specifications

Specifications

Formula Weight 272.18
Flash Point >110°C (230°F)
Molecular Formula C13H5F5O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Boiling Point 93°C (0.2mmHg)
Synonym Pentafluorophenyl phenyl ketone
Beilstein 2059098
CAS 1536-23-8
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzophenone
Melting Point 36° to 39°C
Assay 98%
Quantity 5g
MDL Number MFCD00000294
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.