Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzamide, 99%

$82.97 - $652.65

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentafluorobenzamide
CAS 652-31-3
Molecular Formula C7H2F5NO
Formula Weight 211.09
MDL Number MFCD00007971
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1839722

  SDS
alfa aesar™
A1839722
100g Each for $652.65

AAA1839706

  SDS
alfa aesar™
A1839706
5g Each for $82.97
Description & Specifications

Specifications

Formula Weight 211.09
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzamide
Assay 99%
Melting Point 146° to 149°C
Molecular Formula C7H2F5NO
Quantity 100g
MDL Number MFCD00007971
CAS 652-31-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.