Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentafluoroaniline, 97%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC147082500

 View more versions of this product

Catalog No. AC147082500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Low Melting Solid
Assay ≥96.0%
Formula Weight 183.08
Quantity 250g
Color White to salmon
Density 73g/mL
Melting Point 33° to 38°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula C6F5NH2
Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
Assay Percent Range 97%
Molecular Formula C6H2F5N
CAS 771-60-8
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00007643
Synonym 2,3,4,5,6-Pentafluorobenzenamine
Health Hazard 1 Warning
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
CAS 771-60-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H2F5N
Linear Formula C6F5NH2
Formula Weight 183.08
MDL Number MFCD00007643
Synonym 2,3,4,5,6-Pentafluorobenzenamine
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS & Safety Documents