Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentafluoroaniline, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC147080050

 View more versions of this product

Catalog No. AC147080050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 183.08
Physical Form Crystalline Low Melting Solid
Quantity 5g
Color White to salmon
Melting Point 33° to 38°C
Density 73g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Linear Formula C6F5NH2
Synonym 2,3,4,5,6-Pentafluorobenzenamine
CAS 771-60-8
Health Hazard 1 Warning
Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
Molecular Formula C6H2F5N
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00007643
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

Specifications

Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
CAS 771-60-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H2F5N
Linear Formula C6F5NH2
Formula Weight 183.08
MDL Number MFCD00007643
Synonym 2,3,4,5,6-Pentafluorobenzenamine
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS & Safety Documents