Please login to your online account to display your discounted pricing

2,3,4,5,6-Pentabromobenzyl alcohol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6-Pentabromobenzyl alcohol
Name Note 97%
CAS 79415-41-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H3Br5O
Formula Weight 502.62
MDL Number MFCD00192377
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC344010050

  SDS
thermo fisher scientific
344010050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 502.62
Physical Form Crystalline Powder
Quantity 5g
Color White to light yellow
TSCA TSCA
Beilstein 06, II, 424
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2,3,4,5,6-Pentabromobenzyl alcohol
Assay Percent Range 97%
CAS 79415-41-1
Name Note 97%
MDL Number MFCD00192377
Molecular Formula C7H3Br5O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.