Please login to your online account to display your discounted pricing

2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one, 99%, ACROS Organics™

$149.17 - $149.17

Chemical Identifiers

Chemical Name or Material 2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one
CAS 21306-21-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6Cl6O
Formula Weight 300.77
MDL Number MFCD00019431
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC243950250

  SDS
thermo fisher scientific
AC243950250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC243950050

  SDS
fisher scientific acros
243950050
Glass bottle 5g Each for $149.17
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 300.77
Color Yellow to brownish
Physical Form Crystalline Solid
Quantity 25g
Melting Point 46° to 52°C
Fieser 11,251; 14,174
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one
Assay Percent Range 99%
MDL Number MFCD00019431
CAS Min % 98.5
Packaging Glass bottle
Molecular Formula C6Cl6O
CAS 21306-21-8
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.