Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3,3'-Trichloro-2,4-bipyridine, 98%

Chemical Identifiers

Chemical Name or Material 2',3,3'-Trichloro-2,4-bipyridine
Molecular Formula C10H5Cl3N2
Formula Weight 259.52
MDL Number MFCD09953507
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2043303

  SDS
alfa aesar™
L2043303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL20433MD

  SDS
alfa aesar™
L20433MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 259.52
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2',3,3'-Trichloro-2,4-bipyridine
Assay 98%
Molecular Formula C10H5Cl3N2
MDL Number MFCD09953507
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Quantity 1g
Health Hazard 1 Danger
Melting Point 168° to 170°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.