missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride

$91.30 - $171.18

Chemical Identifiers

CAS 418788-90-6
Molecular Formula C11H16Cl3NO2
Molecular Weight (g/mol) 300.604
MDL Number MFCD06410993
InChI Key GFCJFYSIPRIHKT-UHFFFAOYSA-N
Synonym 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
PubChem CID 52913859
IUPAC Name 2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)Cl)OCCCNCCO.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64981LB0 View Documents Alfa Aesar™
J64981LB0
5mg Each for $91.30
 
AAJ64981MA View Documents Alfa Aesar™
J64981MA
10mg Each for $171.18
 
Specifications

Chemical Identifiers

418788-90-6
300.604
GFCJFYSIPRIHKT-UHFFFAOYSA-N
52913859
C1=CC(=C(C(=C1)Cl)Cl)OCCCNCCO.Cl
C11H16Cl3NO2
MFCD06410993
2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride

Specifications

White
418788-90-6
MFCD06410993
Soluble in water at ≈25 mg/ml; Also soluble in ethanol, DMSO or DMF at ≈20mg/ml; Soluble in PBS, pH 7.2 at ≈10 mg/ml
C1=CC(=C(C(=C1)Cl)Cl)OCCCNCCO.Cl
300.604
300.61
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
5mg
C11H16Cl3NO2
2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
GFCJFYSIPRIHKT-UHFFFAOYSA-N
2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
52913859
Powder
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C