Please login to your online account to display your discounted pricing

2-(2-Tritylaminothiazole-4-yl)-2-methoxyiminoacetic acid hydrochloride, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC247960100

 View more versions of this product

Catalog No. AC247960100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 479.99
Quantity 10g
Color Off-white
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-(2-Tritylaminothiazole-4-yl)-2-methoxyiminoacetic acid hydrochloride
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C25H21N3O3S·HCl
Packaging Glass bottle
MDL Number MFCD00198025
CAS 123333-74-4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(2-Tritylaminothiazole-4-yl)-2-methoxyiminoacetic acid hydrochloride
Name Note 97%
CAS 123333-74-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C25H21N3O3S·HCl
Formula Weight 479.99
MDL Number MFCD00198025
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents