missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Alfa Aesar™

$62.46 - $925.27

Chemical Identifiers

CAS 774-18-5
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD00128232
InChI Key LIERORLYMWHXDL-UHFFFAOYSA-N
Synonym 2-2-trifluoromethyl phenyl ethanamine, o-trifluoromethyl phenethylamine, 2-2-trifluoromethyl phenyl ethan-1-amine, 2-o-trifluoromethylphenyl ethylamine, 2-2-trifluoromethylphenyl ethylamine, 2-trifluoromethyl phenethylamine, 2-2-trifluoromethyl-phenyl-ethylamine, 2-2-trifluoromethyl phenyl ethyl amine, 2-trifluoromethylphenylethanamine, benzeneethanamine,2-trifluoromethyl
PubChem CID 69883
IUPAC Name 2-[2-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC=C(C(=C1)CCN)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6449203 View Documents Alfa Aesar
H6449203
1g Each for $62.46
 
AAH6449206 View Documents Alfa Aesar
H6449206
5g Each for $231.32
 
AAH6449214 View Documents Alfa Aesar
H6449214
25g Each for $925.27
 
Specifications

Chemical Identifiers

774-18-5
189.181
LIERORLYMWHXDL-UHFFFAOYSA-N
69883
C1=CC=C(C(=C1)CCN)C(F)(F)F
C9H10F3N
MFCD00128232
2-2-trifluoromethyl phenyl ethanamine, o-trifluoromethyl phenethylamine, 2-2-trifluoromethyl phenyl ethan-1-amine, 2-o-trifluoromethylphenyl ethylamine, 2-2-trifluoromethylphenyl ethylamine, 2-trifluoromethyl phenethylamine, 2-2-trifluoromethyl-phenyl-ethylamine, 2-2-trifluoromethyl phenyl ethyl amine, 2-trifluoromethylphenylethanamine, benzeneethanamine,2-trifluoromethyl
2-[2-(trifluoromethyl)phenyl]ethanamine

Specifications

Yellow
774-18-5
MFCD00128232
7202211
LIERORLYMWHXDL-UHFFFAOYSA-N
2-[2-(trifluoromethyl)phenyl]ethanamine
69883
Liquid
2-[2-(Trifluoromethyl)phenyl]ethylamine
1g
C9H10F3N
UN2735
2-2-trifluoromethyl phenyl ethanamine, o-trifluoromethyl phenethylamine, 2-2-trifluoromethyl phenyl ethan-1-amine, 2-o-trifluoromethylphenyl ethylamine, 2-2-trifluoromethylphenyl ethylamine, 2-trifluoromethyl phenethylamine, 2-2-trifluoromethyl-phenyl-ethylamine, 2-2-trifluoromethyl phenyl ethyl amine, 2-trifluoromethylphenylethanamine, benzeneethanamine,2-trifluoromethyl
C1=CC=C(C(=C1)CCN)C(F)(F)F
189.181
189.18
95%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.