Learn More
2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6449203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-[2-(Trifluoromethyl)phenyl]ethylamine | |
Yellow | |
MFCD00128232 | |
UN2735 | |
2-2-trifluoromethyl phenyl ethanamine, o-trifluoromethyl phenethylamine, 2-2-trifluoromethyl phenyl ethan-1-amine, 2-o-trifluoromethylphenyl ethylamine, 2-2-trifluoromethylphenyl ethylamine, 2-trifluoromethyl phenethylamine, 2-2-trifluoromethyl-phenyl-ethylamine, 2-2-trifluoromethyl phenyl ethyl amine, 2-trifluoromethylphenylethanamine, benzeneethanamine,2-trifluoromethyl | |
NCCC1=CC=CC=C1C(F)(F)F | |
189.18 | |
189.18 | |
Liquid |
774-18-5 | |
C9H10F3N | |
1 g | |
7202211 | |
LIERORLYMWHXDL-UHFFFAOYSA-N | |
2-[2-(trifluoromethyl)phenyl]ethanamine | |
69883 | |
95% |
Chemical Identifiers
774-18-5 | |
189.18 | |
LIERORLYMWHXDL-UHFFFAOYSA-N | |
69883 | |
NCCC1=CC=CC=C1C(F)(F)F |
C9H10F3N | |
MFCD00128232 | |
2-2-trifluoromethyl phenyl ethanamine, o-trifluoromethyl phenethylamine, 2-2-trifluoromethyl phenyl ethan-1-amine, 2-o-trifluoromethylphenyl ethylamine, 2-2-trifluoromethylphenyl ethylamine, 2-trifluoromethyl phenethylamine, 2-2-trifluoromethyl-phenyl-ethylamine, 2-2-trifluoromethyl phenyl ethyl amine, 2-trifluoromethylphenylethanamine, benzeneethanamine,2-trifluoromethyl | |
2-[2-(trifluoromethyl)phenyl]ethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 212-262-5
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only