missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Thiobisacetamide, 97%

$41.28 - $516.09

Chemical Identifiers

CAS 14618-65-6
Molecular Formula S(CH2CONH2)2
Formula Weight 148.18
MDL Number MFCD00014806
Synonym 2,2'-Thiodiacetamide
Chemical Name or Material 2,2'-Thiobisacetamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2525403 SDS Alfa Aesar™
B2525403
1g Each for $41.28
AAB2525406 SDS Alfa Aesar™
B2525406
5g Each for $135.53
AAB2525414 SDS Alfa Aesar™
B2525414
25g Each for $516.09
Specifications Description & Specifications

Chemical Identifiers

14618-65-6
148.18
2,2'-Thiodiacetamide
S(CH2CONH2)2
MFCD00014806
2,2'-Thiobisacetamide

Specifications

172° to 176°C
14618-65-6
148.18
2,2'-Thiodiacetamide
97%
1g
S(CH2CONH2)2
MFCD00014806
2,2'-Thiobisacetamide