Please login to your online account to display your discounted pricing

2-(2-Thienylsulfonyl)ethanethioamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(2-Thienylsulfonyl)ethanethioamide
CAS 175202-34-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7NO2S3
Formula Weight 221.32
MDL Number MFCD00052277
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM03009CB

  SDS
fisher scientific company
KM03009CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM03009DA

  SDS
fisher scientific company
KM03009DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.32
Quantity 250mg
Assay 97%
CAS Min % 97.0
CAS 175202-34-3
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C6H7NO2S3
Chemical Name or Material 2-(2-Thienylsulfonyl)ethanethioamide
MDL Number MFCD00052277
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.