missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Thienyl)thiazole-4-carboxylic acid, 97%

$43.72 - $596.27

Chemical Identifiers

CAS 24044-07-3
Molecular Formula C8H5NO2S2
Molecular Weight (g/mol) 211.253
MDL Number MFCD01936014
InChI Key FGKCNTGJZXHKFJ-UHFFFAOYSA-N
Synonym 2-2-thienyl-1,3-thiazole-4-carboxylic acid
PubChem CID 2776317
IUPAC Name 2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid
SMILES C1=CSC(=C1)C2=NC(=CS2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54347MD View Documents Alfa Aesar™
H54347MD
250mg Each for $43.72
 
AAH5434703 View Documents Alfa Aesar™
H5434703
1g Each for $138.07
 
AAH5434706 View Documents Alfa Aesar™
H5434706
5g Each for $596.27
 
Specifications

Chemical Identifiers

24044-07-3
211.253
FGKCNTGJZXHKFJ-UHFFFAOYSA-N
2776317
C1=CSC(=C1)C2=NC(=CS2)C(=O)O
C8H5NO2S2
MFCD01936014
2-2-thienyl-1,3-thiazole-4-carboxylic acid
2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid

Specifications

250mg
C8H5NO2S2
2-2-thienyl-1,3-thiazole-4-carboxylic acid
C1=CSC(=C1)C2=NC(=CS2)C(=O)O
211.253
211.25
2-(2-Thienyl)thiazole-4-carboxylic acid
24044-07-3
MFCD01936014
FGKCNTGJZXHKFJ-UHFFFAOYSA-N
2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid
2776317
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures