missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Thienyl)thiazole-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5434706

 View more versions of this product

Catalog No. AAH5434706



Description

Description

Specifications

Specifications

2-(2-Thienyl)thiazole-4-carboxylic acid
24044-07-3
MFCD01936014
FGKCNTGJZXHKFJ-UHFFFAOYSA-N
2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid
2776317
97%
5g
C8H5NO2S2
2-2-thienyl-1,3-thiazole-4-carboxylic acid, 2-2-thienyl-1,3-thiazole-4-carboxylicacid, 2-thien-2-yl-1,3-thiazole-4-carboxylic acid, 2-thiophen-2-yl thiazole-4-carboxylic acid, 2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid, 2-thiophen-2-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2-thienyl
C1=CSC(=C1)C2=NC(=CS2)C(=O)O
211.253
211.25

Chemical Identifiers

24044-07-3
211.253
FGKCNTGJZXHKFJ-UHFFFAOYSA-N
2776317
C1=CSC(=C1)C2=NC(=CS2)C(=O)O
C8H5NO2S2
MFCD01936014
2-2-thienyl-1,3-thiazole-4-carboxylic acid, 2-2-thienyl-1,3-thiazole-4-carboxylicacid, 2-thien-2-yl-1,3-thiazole-4-carboxylic acid, 2-thiophen-2-yl thiazole-4-carboxylic acid, 2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid, 2-thiophen-2-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2-thienyl
2-thiophen-2-yl-1,3-thiazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents