Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Thienyl)pyrrolidine, 97%

$122.05 - $424.91

Chemical Identifiers

Chemical Name or Material 2-(2-Thienyl)pyrrolidine
CAS 90090-64-5
Molecular Formula C8H11NS
Formula Weight 153.25
MDL Number MFCD00052889
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5017903

  SDS
alfa aesar™
H5017903
1g Each for $424.91

AAH50179MD

  SDS
alfa aesar™
H50179MD
250mg Each for $122.05
Description & Specifications

Specifications

Formula Weight 153.25
Chemical Name or Material 2-(2-Thienyl)pyrrolidine
Assay 97%
Molecular Formula C8H11NS
MDL Number MFCD00052889
CAS 90090-64-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
2-(2-Thienyl)pyrrolidine is used in the preparation of imidazopyridine derivatives for treating viral infections.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.