missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Thienyl)pyridine, TCI America™

Manufacturer:  TCI America T293625G

 View more versions of this product

Catalog No. T293625G



Specifications

Specifications

2-(2-Thienyl)pyridine
Yellow
25g
C9H7NS
2-2-thienyl pyridine, 2-thiophen-2-yl pyridine, 2-thien-2-ylpyridine, 2-2-pyridyl thiophene, pyridine, 2-2-thienyl, thienyl pyridine, 2-thienylpyridine, 2-pyrid-2-ylthiophene, acmc-1cknd, 2-thien-2-yl pyridine
C1=CC=NC(=C1)C2=CC=CS2
161.222
161.22
≥98.0% (GC)
140°C
63°C
3319-99-1
MFCD00044441
QLPKTAFPRRIFQX-UHFFFAOYSA-N
2-thiophen-2-ylpyridine
76832
Crystal-Powder at 20°C

Chemical Identifiers

3319-99-1
161.222
QLPKTAFPRRIFQX-UHFFFAOYSA-N
76832
C1=CC=NC(=C1)C2=CC=CS2
C9H7NS
MFCD00044441
2-2-thienyl pyridine, 2-thiophen-2-yl pyridine, 2-thien-2-ylpyridine, 2-2-pyridyl thiophene, pyridine, 2-2-thienyl, thienyl pyridine, 2-thienylpyridine, 2-pyrid-2-ylthiophene, acmc-1cknd, 2-thien-2-yl pyridine
2-thiophen-2-ylpyridine
Safety and Handling

Safety and Handling

TSCA : No

Documents