Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Thienyl)piperazine, 96%

$123.24 - $313.41

Chemical Identifiers

Chemical Name or Material 2-(2-Thienyl)piperazine
CAS 85803-49-2
Molecular Formula C8H12N2S
Formula Weight 168.26
MDL Number MFCD06245415
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2673303

  SDS
alfa aesar™
H2673303
1g Each for $313.41

AAH26733MD

  SDS
alfa aesar™
H26733MD
250mg Each for $123.24
Description & Specifications

Specifications

Formula Weight 168.26
Melting Point 82° to 84°C
Quantity 1g
Assay 96%
Molecular Formula C8H12N2S
MDL Number MFCD06245415
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 85803-49-2
Chemical Name or Material 2-(2-Thienyl)piperazine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.