Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Thenoin, 97%

$72.80 - $938.96

Chemical Identifiers

Chemical Name or Material 2,2'-Thenoin
CAS 27761-02-0
Molecular Formula C10H8O2S2
Formula Weight 224.29
MDL Number MFCD00022505
Synonym 2-Hydroxy-1,2-di-2-thienylethanone; alpha-Hydroxy 2-thenyl 2-thienyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2467103

 
alfa aesar™
B2467103
1g Each for $72.80

AAB2467106

 
alfa aesar™
B2467106
5g Each for $243.20

AAB2467114

 
alfa aesar™
B2467114
25g Each for $938.96
Description & Specifications

Specifications

Formula Weight 224.29
Chemical Name or Material 2,2'-Thenoin
Assay 97%
Molecular Formula C10H8O2S2
MDL Number MFCD00022505
CAS 27761-02-0
Synonym 2-Hydroxy-1,2-di-2-thienylethanone; alpha-Hydroxy 2-thenyl 2-thienyl ketone
Melting Point 104° to 107°C
Quantity 1g