missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-Thenoin, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals B2467103
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2,2'-Thenoin | |
104°C to 107°C | |
MFCD00022505 | |
2,2'-thenoin, 2-hydroxy-1,2-di-2-thienylethanone, 2-hydroxy-1,2-di 2-thienyl ethan-1-one, 2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one, 2-hydroxy-1,2-di thiophen-2-yl ethanone, 2-hydroxy-1,2-bis thiophen-2-yl ethanone, maybridge1_002119, 1,2-dithien-2-ylhydroxyethanone, 2-hydroxy-1,2-dithien-2-ylethanone, alpha-hydroxy-2-thenyl 2-thienyl ketone | |
C1=CSC(=C1)C(C(=O)C2=CC=CS2)O | |
224.292 | |
224.29 |
27761-02-0 | |
C10H8O2S2 | |
1 g | |
OGWZIOZTPNLTCR-UHFFFAOYSA-N | |
2-hydroxy-1,2-dithiophen-2-ylethanone | |
2734800 | |
97% |
Chemical Identifiers
27761-02-0 | |
224.292 | |
OGWZIOZTPNLTCR-UHFFFAOYSA-N | |
2734800 | |
C1=CSC(=C1)C(C(=O)C2=CC=CS2)O |
C10H8O2S2 | |
MFCD00022505 | |
2,2'-thenoin, 2-hydroxy-1,2-di-2-thienylethanone, 2-hydroxy-1,2-di 2-thienyl ethan-1-one, 2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one, 2-hydroxy-1,2-di thiophen-2-yl ethanone, 2-hydroxy-1,2-bis thiophen-2-yl ethanone, maybridge1_002119, 1,2-dithien-2-ylhydroxyethanone, 2-hydroxy-1,2-dithien-2-ylethanone, alpha-hydroxy-2-thenyl 2-thienyl ketone | |
2-hydroxy-1,2-dithiophen-2-ylethanone |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures