missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Pyridyl)imidazole-4,5-dithiocarboxamide, 97%

Chemical Identifiers

CAS 1347814-87-2
Molecular Formula C10H9N5S2
Molecular Weight (g/mol) 263.337
MDL Number MFCD20265252
InChI Key QDQGYOFBSXUZFH-UHFFFAOYSA-N
Synonym 2-2-pyridyl imidazole-4,5-dithiocarboxamide
PubChem CID 73995777
IUPAC Name 2-pyridin-2-yl-1H-imidazole-4,5-dicarbothioamide
SMILES C1=CC=NC(=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54368MD View Documents Alfa Aesar™
H54368MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5436803 View Documents Alfa Aesar™
H5436803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5436806 View Documents Alfa Aesar™
H5436806
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1347814-87-2
263.337
QDQGYOFBSXUZFH-UHFFFAOYSA-N
73995777
C1=CC=NC(=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
C10H9N5S2
MFCD20265252
2-2-pyridyl imidazole-4,5-dithiocarboxamide
2-pyridin-2-yl-1H-imidazole-4,5-dicarbothioamide

Specifications

250mg
C10H9N5S2
2-2-pyridyl imidazole-4,5-dithiocarboxamide
C1=CC=NC(=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
263.337
263.33
2-(2-Pyridyl)imidazole-4,5-dithiocarboxamide
1347814-87-2
MFCD20265252
QDQGYOFBSXUZFH-UHFFFAOYSA-N
2-pyridin-2-yl-1H-imidazole-4,5-dicarbothioamide
73995777
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures