missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Pyridyl)benzimidazole, TCI America™

Manufacturer:  TCI America P15995G

 View more versions of this product

Catalog No. P15995G



Specifications

Specifications

2-(2-Pyridyl)benzimidazole
225°C
1137-68-4
MFCD00005586
YNFBMDWHEHETJW-UHFFFAOYSA-N
2-pyridin-2-yl-1H-benzimidazole
70821
Crystal-Powder at 20°C
Undesignated
5g
C12H9N3
2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzo d imidazole, 2-pyridin-2-yl-1h-benzoimidazole, 2-pyridin-2-yl-1h-1,3-benzodiazole, 2-pyridin-2-yl-1h-benzimidazole, chembl72683, 1h-benzimidazole, 2-pyridinyl, 1h-benzimidazole, 2-2-pyridinyl, acmc-20aime, 2 2-pyridyl benzimidazole
C1=CC=C2C(=C1)NC(=N2)C3=CC=CC=N3
195.225
195.23
≥98.0% (HPLC,T)

Chemical Identifiers

1137-68-4
195.225
YNFBMDWHEHETJW-UHFFFAOYSA-N
70821
C1=CC=C2C(=C1)NC(=N2)C3=CC=CC=N3
C12H9N3
MFCD00005586
2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzo d imidazole, 2-pyridin-2-yl-1h-benzoimidazole, 2-pyridin-2-yl-1h-1,3-benzodiazole, 2-pyridin-2-yl-1h-benzimidazole, chembl72683, 1h-benzimidazole, 2-pyridinyl, 1h-benzimidazole, 2-2-pyridinyl, acmc-20aime, 2 2-pyridyl benzimidazole
2-pyridin-2-yl-1H-benzimidazole
Safety and Handling

Safety and Handling

TSCA : No

Documents