Please login to your online account to display your discounted pricing

2-(2-Pyridyl)benzimidazole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(2-Pyridyl)benzimidazole
Name Note 99%
CAS 1137-68-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H9N3
Formula Weight 195.22
MDL Number MFCD00005586
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC167950050

  SDS
fisher scientific acros
167950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC167950100

  SDS
fisher scientific acros
167950100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 195.22
Quantity 5g
Color Yellow to brown
EINECS Number 214-508-7
TSCA TSCA
Melting Point 220° to 225°C
CAS Max % 100.0
Chemical Name or Material 2-(2-Pyridyl)benzimidazole
Packaging Glass bottle
Molecular Formula C12H9N3
MDL Number MFCD00005586
CAS Min % 98.5
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
CAS 1137-68-4