Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Pyridyl)benzimidazole, 98+%

$28.93 - $28.93

Chemical Identifiers

Chemical Name or Material 2-(2-Pyridyl)benzimidazole
CAS 1137-68-4
Molecular Formula C12H9N3
Formula Weight 195.22
MDL Number MFCD00005586
Beilstein 151411
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0133903

 
alfa aesar™
L0133903
1g Each for $28.93
Description & Specifications

Specifications

Formula Weight 195.22
Assay ≥98%
MDL Number MFCD00005586
CAS 1137-68-4
Chemical Name or Material 2-(2-Pyridyl)benzimidazole
Molecular Formula C12H9N3
Beilstein 151411
Melting Point 221° to 225°C
Quantity 1g

Applications
2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.