missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Propynyloxy)ethylamine 98.0+%, TCI America™
$608.86 - $3034.32
Chemical Identifiers
CAS | 122116-12-5 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.133 |
InChI Key | MZFRRBQTCNHISM-UHFFFAOYSA-N |
PubChem CID | 18356755 |
IUPAC Name | 2-prop-2-ynoxyethanamine |
SMILES | C#CCOCCN |
Chemical Identifiers
122116-12-5 | |
99.133 | |
18356755 | |
C#CCOCCN |
C5H9NO | |
MZFRRBQTCNHISM-UHFFFAOYSA-N | |
2-prop-2-ynoxyethanamine |
Specifications
122116-12-5 | |
C5H9NO | |
2734 | |
C#CCOCCN | |
99.133 | |
99.13 | |
Liquid |
Yellow | |
1 g | |
MZFRRBQTCNHISM-UHFFFAOYSA-N | |
2-prop-2-ynoxyethanamine | |
18356755 | |
≥98.0% (T) | |
2-(2-Propynyloxy)ethylamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator