Please login to your online account to display your discounted pricing

2-[(2-Phenoxyethyl)thio]acetic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM06649EA

 View more versions of this product

Catalog No. KM06649EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 212.27
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-[(2-Phenoxyethyl)thio]acetic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H12O3S
Linear Formula C6H5O(CH2)2SCH2CO2H
MDL Number MFCD00085152
CAS 75434-70-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2-[(2-Phenoxyethyl)thio]acetic acid
CAS 75434-70-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12O3S
Linear Formula C6H5O(CH2)2SCH2CO2H
Formula Weight 212.27
MDL Number MFCD00085152
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents