missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-Oxybis(ethylamine), 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 275840010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 2'-Oxybis(ethylamine) | |
Colorless | |
69.0°C to 71.0°C (15.0 mmHg) | |
97.5% min. (GC) | |
C4H12N2O | |
NH2CH2CH2OCH2CH2NH2 | |
1 g | |
0.961 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform | |
NCCOCCN | |
104.15 | |
104.15 | |
Liquid |
2752-17-2 | |
0.9610g/mL | |
Authentic | |
Glass bottle | |
1.4550 to 1.4580 | |
MFCD00466591 | |
04,IV,1414 | |
2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine | |
GXVUZYLYWKWJIM-UHFFFAOYSA-N | |
2-(2-aminoethoxy)ethanamine | |
75982 | |
98% |
Chemical Identifiers
2752-17-2 | |
104.15 | |
GXVUZYLYWKWJIM-UHFFFAOYSA-N | |
75982 | |
NCCOCCN |
C4H12N2O | |
MFCD00466591 | |
2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine | |
2-(2-aminoethoxy)ethanamine |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 220-395-5
RUO – Research Use Only