missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-Oxybis(ethylamine), 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC275840050
Specifications
2, 2'-Oxybis(ethylamine) | |
Authentic | |
1.4550 to 1.4580 | |
69.0°C to 71.0°C (15.0mmHg) | |
5g | |
97.5% min. (GC) | |
NH2CH2CH2OCH2CH2NH2 | |
1,5-diamino-3-oxapentane, 2,2'-diaminodiethyl ether, 2,2'-oxybis ethylamine, 2,2'-oxydiethanamine, 2,2'-oxydiethylamine, 2-2-aminoethoxy ethanamine, aminoethyl ether, bis 2-aminoethyl ether, ethanamine, 2,2'-oxybis, unii-ffw1773zz6 | |
GXVUZYLYWKWJIM-UHFFFAOYSA-N | |
2-(2-aminoethoxy)ethanamine | |
75982 | |
Liquid |
0.9610g/mL | |
Glass bottle | |
0.961 | |
Colorless | |
2752-17-2 | |
C4H12N2O | |
04,IV,1414 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform | |
C(COCCN)N | |
104.15 | |
104.15 | |
98% |
Chemical Identifiers
2752-17-2 | |
104.15 | |
1,5-diamino-3-oxapentane, 2,2'-diaminodiethyl ether, 2,2'-oxybis ethylamine, 2,2'-oxydiethanamine, 2,2'-oxydiethylamine, 2-2-aminoethoxy ethanamine, aminoethyl ether, bis 2-aminoethyl ether, ethanamine, 2,2'-oxybis, unii-ffw1773zz6 | |
2-(2-aminoethoxy)ethanamine |
C4H12N2O | |
GXVUZYLYWKWJIM-UHFFFAOYSA-N | |
75982 | |
C(COCCN)N |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 220-395-5