missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Nitroanilino)ethanol, TCI America™

$189.45 - $660.02

Chemical Identifiers

CAS 4926-55-0
Molecular Formula C8H10N2O3
Molecular Weight (g/mol) 182.179
MDL Number MFCD00239475
InChI Key LFOUYKNCQNVIGI-UHFFFAOYSA-N
Synonym N-(2-Hydroxyethyl)-2-nitroaniline, N-(2-Nitrophenyl)ethanolamine
PubChem CID 78637
IUPAC Name 2-(2-nitroanilino)ethanol
SMILES C1=CC=C(C(=C1)NCCO)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N100025G View Documents TCI America
N100025G
25g Each for $660.02
 
N10005G View Documents TCI America
N10005G
5g Each for $189.45
 
Specifications

Chemical Identifiers

4926-55-0
182.179
LFOUYKNCQNVIGI-UHFFFAOYSA-N
78637
C1=CC=C(C(=C1)NCCO)[N+](=O)[O-]
C8H10N2O3
MFCD00239475
N-(2-Hydroxyethyl)-2-nitroaniline, N-(2-Nitrophenyl)ethanolamine
2-(2-nitroanilino)ethanol

Specifications

Undesignated
25g
C8H10N2O3
N-(2-Hydroxyethyl)-2-nitroaniline, N-(2-Nitrophenyl)ethanolamine
C1=CC=C(C(=C1)NCCO)[N+](=O)[O-]
182.179
182.18
≥98.0% (GC)
73°C
4926-55-0
MFCD00239475
LFOUYKNCQNVIGI-UHFFFAOYSA-N
2-(2-nitroanilino)ethanol
78637
Crystal-Powder at 20°C
2-(2-Nitroanilino)ethanol
Safety and Handling

Safety and Handling

TSCA : Yes