Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Naphthyl)indole, 98%

$111.59 - $431.00

Chemical Identifiers

Chemical Name or Material 2-(2-Naphthyl)indole
CAS 23746-81-8
Molecular Formula C18H13N
Formula Weight 243.31
MDL Number MFCD00124144
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2043403

  SDS
alfa aesar™
B2043403
1g Each for $111.59

AAB2043406

  SDS
alfa aesar™
B2043406
5g Each for $431.00
Description & Specifications

Specifications

Formula Weight 243.31
Chemical Name or Material 2-(2-Naphthyl)indole
Assay 98%
Molecular Formula C18H13N
MDL Number MFCD00124144
CAS 23746-81-8
Quantity 1g

Applications
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

Solubility
Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.