Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Naphthoyl)-1-(p-toluoyl)hydrazine, 98%

$40.23 - $40.23

Chemical Identifiers

Chemical Name or Material 2-(2-Naphthoyl)-1-(p-toluoyl)hydrazine
CAS 83803-95-6
Molecular Formula C19H16N2O2
Formula Weight 304.33
MDL Number MFCD00021629
Synonym 1-(4-Methylbenzoyl)-2-(2-naphthoyl)hydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1859003

  SDS
alfa aesar™
A1859003
1g Each for $40.23
Description & Specifications

Specifications

Formula Weight 304.33
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 83803-95-6
Synonym 1-(4-Methylbenzoyl)-2-(2-naphthoyl)hydrazine
Molecular Formula C19H16N2O2
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD00021629
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(2-Naphthoyl)-1-(p-toluoyl)hydrazine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.